EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry frontiers
Rapid Synthesis of Redox-Active Dodecaborane B
PMID 27885335
Abstract
We have developed a fast and efficient route to obtain perfunctionalized ether-linked alkyl and benzyl derivatives of the
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
901272
Dodeca(2,3,4,5,6-pentafluorobenzyloxy)-hypercloso-dodecaborane
C84H24B12F60O12