EMAIL THIS PAGE TO A FRIEND
Liver international : official journal of the International Association for the Study of the Liver
17,β-estradiol inhibits hepatitis C virus mainly by interference with the release phase of its life cycle.
PMID 27885811
Abstract
Oestrogen and oestrogen-mediated signalling protect from hepatitis C virus through incompletely understood mechanisms. We aimed to ascertain which phase(s) of hepatitis C virus life cycle is/are affected by oestrogens. Huh7 cells infected with the JFH1 virus (genotype 2a) were exposed to dehydroepiandrosterone, testosterone, progesterone and 17β-estradiol (tested with/without its receptor antagonist fulvestrant). Dose-response curves were established to calculate half maximal inhibitory concentration values. To dissect how 17β-estradiol interferes with phases of hepatitis C virus life cycle, its effects were measured on the hepatitis C virus pseudo-particle system (viral entry), the subgenomic replicon N17/JFH1 and the replicon cell line Huh7-J17 (viral replication). Finally, in a dual-step infection model, infectious supernatants, collected from infected cells exposed to hormones, were used to infect naïve cells. Progesterone and testosterone showed no inhibitory effect on hepatitis C virus; dehydroepiandrosterone was only mildly inhibitory. In contrast, 17β-estradiol inhibited infection by 64%-67% (IC 17β-estradiol inhibits hepatitis C virus acting through its intracellular receptors, mainly interfering with late phases (assembly/release) of the hepatitis C virus life cycle.