EMAIL THIS PAGE TO A FRIEND
Transgenic research
A novel heavy metal ATPase peptide from Prosopis juliflora is involved in metal uptake in yeast and tobacco.
PMID 27888434
Abstract
Heavy metal pollution of agricultural soils is one of the most severe ecological problems in the world. Prosopis juliflora, a phreatophytic tree species, grows well in heavy metal laden industrial sites and is known to accumulate heavy metals. Heavy Metal ATPases (HMAs) are ATP driven heavy metal pumps that translocate heavy metals across biological membranes thus helping the plant in heavy metal tolerance and phytoremediation. In the present study we have isolated and characterized a novel 28.9 kDa heavy metal ATPase peptide (PjHMT) from P. juliflora which shows high similarity to the C-terminal region of P
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8640
E-64d, protease inhibitor
C17H30N2O5