EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Label-free piezoelectric biosensor for prognosis and diagnosis of Systemic Lupus Erythematosus.
PMID 27888685
Abstract
An autoantigen piezoelectric sensor to quantify specific circulating autoantibodies in human serum is developed. The sensor consisted on a quartz crystal microbalance with dissipation monitoring (QCM-D) where TRIM21 and TROVE2 autoantigens were covalently immobilized, allowing the selective determination of autoantibodies for diagnosis and prognosis of Systemic Lupus Erythematosus (SLE). The sensitivity of the biosensor, measured as IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C11006
Carbohydrazide, 98%
CH6N4O