EMAIL THIS PAGE TO A FRIEND
Oncotarget
Exosomal long noncoding RNA CRNDE-h as a novel serum-based biomarker for diagnosis and prognosis of colorectal cancer.
PMID 27888803
Abstract
Cancer-secreted long non-coding RNAs (lncRNAs) are emerging mediators of cancer-host cross talk. The aim of our study was to illustrate the clinical significance of the lncRNA CRNDE-h in exosomes purified from the serum of patients with colorectal cancer (CRC). The study was divided into four parts: (1) The exosome isolated methods and lncRNA detected methods which accurately and reproducibly measure CRC-related exosomal CRNDE-h in serum were optimized in preliminary pilot stage; (2) The stability of exosomal CRNDE-h was evaluated systematically; (3) The origin of exosomal CRNDE-h was explorated in vitro and in vivo; (4) The diagnostic and prognostic value of exosomal CRNDE-h for CRC were validated in 468 patients. In pilot study, our results indicated that exosomal CRNDE-h was detectable and stable in serum of CRC patients, and derived from tumor cells. Then, the increased expression of exosomal CRNDE-h was successfully validated in 148 CRC patients when compared with colorectal benign disease patients and healthy donors. Exosomal CRNDE-h level significantly correlated with CRC regional lymph node metastasis (P = 0.019) and distant metastasis (P = 0.003). Moreover, at the cut-off value of 0.020 exosomal CRNDE-h level of serum, the area under ROC curve distinguishing CRC from colorectal benign disease patients and healthy donors was 0.892, with 70.3% sensitivity and 94.4% specificity, which was superior to carcinoembryogenic antigen. In addition, high exosomal CRNDE-h level has a lower overall survival rates than that for low groups (34.6% vs. 68.2%, P < 0.001). In conclusion, detection of lncRNA CRNDE-h in exosome shed a light on utilizing exosomal CRNDE-h as a noninvasive serum-based tumor marker for diagnosis and prognosis of CRC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
729817
2-Chloroethyl acrylate, contains >100 ppm MEHQ as inhibitor, 97%
C5H7ClO2