EMAIL THIS PAGE TO A FRIEND
Chemosphere
By-product identification and phytotoxicity of biodegraded Direct Yellow 4 dye.
PMID 27889513
Abstract
Citrus limon peroxidase mediated decolourization of Direct Yellow 4 (DY4) was investigated. The process variables (pH, temperature, incubation time, enzyme dose, H