EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Spectroscopic investigation and direct comparison of the reactivities of iron pyridyl oxidation catalysts.
PMID 27889672
Abstract
The growing interest in green chemistry has fueled attention to the development and characterization of effective iron complex oxidation catalysts. A number of iron complexes are known to catalyze the oxidation of organic substrates utilizing peroxides as the oxidant. Their development is complicated by a lack of direct comparison of the reactivities of the iron complexes. To begin to correlate reactivity with structural elements, we compare the reactivities of a series of iron pyridyl complexes toward a single dye substrate, malachite green (MG), for which colorless oxidation products are established. Complexes with tetradentate, nitrogen-based ligands with cis open coordination sites were found to be the most reactive. While some complexes reflect sensitivity to different peroxides, others are similarly reactive with either H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
38800
Malachite Green chloride, analytical standard
C23H25ClN2