EMAIL THIS PAGE TO A FRIEND
Travel medicine and infectious disease
Investigation of carbapenem resistance and the first identification of Klebsiella pneumoniae carbapenemase (KPC) enzyme among Escherichia coli isolates in Turkey: A prospective study.
PMID 27890667
Abstract
The aim of this study was to determine the presence of carbapenem resistance and carbapenemase production in Escherichia coli isolates from clinical samples in Turkey. The prospective study included a total of 4.052 Escherichia coli isolates collected from patients admitted to a hospital from March 2011 to May 2012. We used ertapenem disc for screening carbapenemase production, and the confirmation was performed by using Etest. The resistance mechanisms and genetic relatedness of the carbapenem resistant strains were investigated by using PCR (polymerase chain reaction) and pulsed-field gel electrophoresis (PFGE), respectively. Among the 4.052 E. coli isolates, 24 (0.59%) were found to be carbapenem resistant. Of these, only 5 isolates were positive for OXA-48 and 2 isolates were positive for Klebsiella pneumoniae carbapenemase (KPC)-2. The KPC-2 producing E. coli strains (n = 2) were both isolated from the same patient. The bla This article confirms, to our knowledge for the first time, the detection of KPC-2-producing E. coli in Turkey, with OXA-48 being the most frequent carbapenemase in the study.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS022172
Ertapenem
C22H25N3O7S