PloS one
Quantification of Arachidonic Acid and Its Metabolites in Rat Tissues by UHPLC-MS/MS: Application for the Identification of Potential Biomarkers of Benign Prostatic Hyperplasia.
PMID 27893755
Abstract
To evaluate the potential relationship between benign prostatic hyperplasia (BPH) and the arachidonic acid (AA) metabolome, a UHPLC-MS/MS method has been developed and validated for simultaneous determination of AA and its cyclooxygenase(COX) and lipoxygenase(LOX) pathway metabolites (15-HETE, 12-HETE, TXA2, 5-HETE, AA, PGI2, PGF2α, 8-HETE, PGD2, PGE2 and LTB4) in rat tissues. The analytes were extracted from tissue samples with a protein precipitation procedure and then separated on a Shim-pack XR-ODSC18 column with 0.05% formic acid in water (pH adjusted with dilute ammonia) and methanol:acetonitrile (20:80, v/v). Detection was performed on a UHPLC-MS/MS system with electrospray negative ionization (ESI) and a multiple reaction-monitoring mode. The lower limits of quantification (LLOQ) were 0.25-50 ng/mL for all of the analytes in the prostate, seminal, bladder, liver and kidney tissues. The absolute recoveries of the analytes from all of the tissues were more than 50%. By means of the method developed, the AA metabolites in tissue samples from Sham and BPH group rats were determined. The eleven biomarkers in the BPH group prostate, seminal, bladder, liver and kidney tissues were significantly higher than those of the sham group, indicating that BPH fortified the inducible expression of COX and LOX, as well as increased the production of AA and eicosanoids. The method described here offers a useful tool for the evaluation of complex regulatory eicosanoids responses in vivo.