EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Human and veterinary pharmaceuticals in the marine environment including fish farms in Korea.
PMID 27894804
Abstract
The occurrence trends and effects of 30 human and veterinary pharmaceuticals, including antibiotics, anthelmintics, anti-inflammatory drugs, and β-blockers, in the marine environment, with a focus on seawater, sediment, cultured fish, and their feed collected from coastal and fish farm areas in the southern sea of Korea, were investigated. The concentrations of total pharmaceuticals in coastal area seawater (mean: 533ng/L) were higher than those in fish farm seawater (mean: 300ng/L), while the opposite trend (coastal area: 136ng/gdrywt
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32567
Fenbendazole-d3, VETRANAL®, analytical standard
C15D3H10N3O2S
34058
Norfloxacin-d5, VETRANAL®, analytical standard
C16H13D5FN3O3