EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Correctors and Potentiators Rescue Function of the Truncated W1282X-Cystic Fibrosis Transmembrane Regulator (CFTR) Translation Product.
PMID 27895116
Abstract
W1282X is the fifth most common cystic fibrosis transmembrane regulator (CFTR) mutation that causes cystic fibrosis. Here, we investigated the utility of a small molecule corrector/potentiator strategy, as used for ΔF508-CFTR, to produce functional rescue of the truncated translation product of the W1282X mutation, CFTR
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3129
N-Octanoyl-N-methylglucamine, ≥97%
C15H31NO6