EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Upconversion nanoparticles grafted molybdenum disulfide nanosheets platform for microcystin-LR sensing.
PMID 27898377
Abstract
Water safety is one of the most pervasive problems afflicting people throughout the world. Microcystin-LR (MC-LR), a representative toxin released by cyanobacteria, poses an increasing and serious threat to water safety. In order to develop facile, specific and sensitive detection methods for MC-LR, we fabricated an ultrasensitive fluorescence aptasensor using the enhanced fluorescence of UCNP and the effective quenching ability, high affinity toward single strand DNA (ssDNA) of MoS