EMAIL THIS PAGE TO A FRIEND
Journal of environmental quality
Persistence in and Release of 2,4-D and Azoxystrobin from Turfgrass Clippings.
PMID 27898773
Abstract
Research has shown that pesticide residue in clippings from previously treated turfgrass may become bioavailable as grass decomposes, adversely affecting off-target organisms. We conducted a field study to quantify 2,4-D (2,4-dichlorophenoxyacetic acid) and azoxystrobin (methyl(E)-2-{2[6-(2-cyanophenoxy)pyrmidin-4-yloxy]phenyl}-3-methoxyacrylate) residues in turfgrass clippings collected from hybrid bermudagrass [ (L.) Pers. × Burtt-Davy], tall fescue [ (Schreb.) S.J. Darbyshire], and zoysiagrass ( Steud.). A subsequent greenhouse experiment was conducted to measure pesticide release from clippings into water. 2,4-D (1.6 kg a.i. ha) and azoxystrobin (0.6 kg a.i. ha) were applied to field plots at 32, 16, 8, 4, 2, 1, or 0 d before collection of the clippings. Clippings were collected from each experimental unit to quantify pesticide release from clippings into water. Both 2,4-D and azoxystrobin were detected when turfgrass was treated over the 32-d experimental period, suggesting that clipping management should be implemented for an extended period of time after application. Pesticide residue was detected in all water samples collected, confirming 2,4-D and azoxystrobin release from turfgrass clippings; however, pesticide release varied between compounds. Two days after clippings were incorporated in water, 39 and 10% of 2,4-D and azoxystrobin were released from clippings, respectively. Our research supports the currently recommended practice of returning clippings to the turfgrass stand when mowing because removal of 2,4-D and azoxystrobin in clippings may reduce pest control and cause adverse off-target impacts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31697
Azoxystrobin, PESTANAL®, analytical standard
C22H17N3O5