Chembiochem : a European journal of chemical biology
Peptoid Efficacy against Polymicrobial Biofilms Determined by Using Propidium Monoazide-Modified Quantitative PCR.
PMID 27900840
Biofilms containing Candida albicans are responsible for a wide variety of clinical infections. The protective effects of the biofilm matrix, the low metabolic activity of microorganisms within a biofilm and their high mutation rate, significantly enhance the resistance of biofilms to conventional antimicrobial treatments. Peptoids are peptide-mimics that share many features of host defence antimicrobial peptides but have increased resistance to proteases and therefore have better stability in vivo. The activity of a library of peptoids was tested against monospecies and polymicrobial bacterial/fungal biofilms. Selected peptoids showed significant bactericidal and fungicidal activity against the polymicrobial biofilms. This coupled with low cytotoxicity suggests that peptoids could offer a new option for the treatment of clinically relevant polymicrobial infections.