EMAIL THIS PAGE TO A FRIEND
Journal of neuroinflammation
Cell cycle inhibition reduces inflammatory responses, neuronal loss, and cognitive deficits induced by hypobaria exposure following traumatic brain injury.
PMID 27903275
Abstract
Traumatic brain injury (TBI) patients in military settings can be exposed to prolonged periods of hypobaria (HB) during aeromedical evacuation. Hypobaric exposure, even with supplemental oxygen to prevent hypoxia, worsens outcome after experimental TBI, in part by increasing neuroinflammation. Cell cycle activation (CCA) after TBI has been implicated as a mechanism contributing to both post-traumatic cell death and neuroinflammation. Here, we examined whether hypobaric exposure in rats subjected to TBI increases CCA and microglial activation in the brain, as compared to TBI alone, and to evaluate the ability of a cyclin-dependent kinase (CDK) inhibitor (CR8) to reduce such changes and improve behavioral outcomes. Adult male Sprague Dawley rats were subjected to fluid percussion-induced injury, and HB exposure was performed at 6 h after TBI. Western blot and immunohistochemistry (IHC) were used to assess cell cycle-related protein expression and inflammation at 1 and 30 days after injury. CR8 was administered intraperitoneally at 3 h post-injury; chronic functional recovery and histological changes were assessed. Post-traumatic hypobaric exposure increased upregulation of cell cycle-related proteins (cyclin D1, proliferating cell nuclear antigen, and CDK4) and microglial/macrophage activation in the ipsilateral cortex at day 1 post-injury as compared to TBI alone. Increased immunoreactivity of cell cycle proteins, as well as numbers of Iba-1 HB exposure following TBI increases CCA, neuroinflammation, and associated neuronal cell loss. These changes and post-traumatic cognitive deficits are reduced by CDK inhibition; such drugs may therefore serve to protect TBI patients requiring aeromedical evacuation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3249
CR8, ≥95% (HPLC)
C24H29N7O