EMAIL THIS PAGE TO A FRIEND
Cell death & disease
1-Methyl-tryptophan attenuates regulatory T cells differentiation due to the inhibition of estrogen-IDO1-MRC2 axis in endometriosis.
PMID 27906184
Abstract
Foxp3
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU003351 MISSION® esiRNA, esiRNA targeting human MRC2 (esiRNA1)