EMAIL THIS PAGE TO A FRIEND
Nanoscale
Unusual reactivity of dithiol protected clusters in comparison to monothiol protected clusters: studies using Ag
PMID 27906399
Abstract
We report the synthesis and unique reactivity of a new green dithiol protected cluster (DTPC), Ag
Related Materials