EMAIL THIS PAGE TO A FRIEND
Antiviral research
Assessment of the potential for host-targeted iminosugars UV-4 and UV-5 activity against filovirus infections in vitro and in vivo.
PMID 27908828
Abstract
Iminosugars are host-directed antivirals with broad-spectrum activity. The iminosugar, N-butyl-deoxynojirimycin (NB-DNJ or Miglustat
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55222
6-Carboxytetramethylrhodamine, BioReagent, suitable for fluorescence, ≥90% (HPLC)
C25H22N2O5