EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Effects of Melatonin, Aluminum Oxide, and Polymethylsiloxane Complex on the Expression of LYVE-1 in the Liver of Mice with Obesity and Type 2 Diabetes Mellitus.
PMID 27909960
Abstract
The effects of melatonin, aluminum oxide, and polymethylsiloxane complex on the expression of LYVE-1 (lymphatic vessel endothelial hyaluronan receptor) in the liver were studied in db/db mice with experimental obesity and type 2 diabetes mellitus. The complex or placebo was administered daily by gavage from week 8 to week 16 of life. The animals receiving the complex exhibited enhanced, in comparison with the placebo group, immunohistochemical LYVE-1+ staining of endothelial cells in sinusoids. Enhanced expression of LYVE-1 was associated with less pronounced dilatation of interlobular arteries, veins, and lymphatic vessels. Thee findings suggest a protective effect of the complex towards structural changes in the liver of mice with obesity and type 2 diabetes.