EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Suppression of NADPH oxidase attenuates hypoxia-induced dysfunctions of endothelial progenitor cells.
PMID 27913300
Abstract
NADPH oxidases (NOX) - derived reactive oxygen species (ROS) contribute to oxidative injury in hypoxia-induced pulmonary arterial hypertension. This study aims to evaluate the status of NOX in endothelial progenitor cells (EPCs) under hypoxic condition and to determine whether NOX inhibitors could attenuate hypoxia-induced dysfunctions of EPCs. EPCs were isolated from peripheral blood of SD rats and subjected to hypoxia (O