EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Lung cellular and molecular physiology
CD36 mediates H2O2-induced calcium influx in lung microvascular endothelial cells.
PMID 27913425
Abstract
Elevated levels of reactive oxygen species and intracellular Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2148
Sulfo-N-succinimidyl Oleate sodium, ≥95% (HPLC)
C22H36NO7S· Na