EMAIL THIS PAGE TO A FRIEND
Virus research
Valosin-containing protein (VCP/p97) plays a role in the replication of West Nile virus.
PMID 27914931
Abstract
Valosin-containing protein (VCP) is classified as a member of the type II AAA
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU071991 MISSION® esiRNA, esiRNA human VCP (esiRNA1)