EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
Multifunctions of dual Zn/Mg ion co-implanted titanium on osteogenesis, angiogenesis and bacteria inhibition for dental implants.
PMID 27915020
Abstract
In order to improve the osseointegration and long-term survival of dental implants, it is urgent to develop a multifunctional titanium surface which would simultaneously have osteogeneic, angiogeneic and antibacterial properties. In this study, a potential dental implant material-dual Zn/Mg ion co-implanted titanium (Zn/Mg-PIII) was developed via plasma immersion ion implantation (PIII). The Zn/Mg-PIII surfaces were found to promote initial adhesion and spreading of rat bone marrow mesenchymal stem cells (rBMSCs) via the upregulation of the gene expression of integrin α1 and integrin β1. More importantly, it was revealed that Zn/Mg-PIII could increase Zn In order to enhance the rapid osseointegration and long-term survival of dental implants, various works on titanium surface modification have been carried out. However, only improving osteogenic activity of implants is not enough, because angiogenesis and bacteria inhibition are also very important for dental implants. In the present study, a novel dental implant material-dual Zn/Mg ion co-implanted titanium (Zn/Mg-PIII) was developed, which was found to have superior osteoinductivity, pro-angiogenic effects and inhibitory effects against oral anaerobes. Furthermore, synergistic effects of Zn/Mg ions on osteogenic differentiation of rBMSCs and the possible mechanism were discovered. In addition, rapid osseointegration and sustained biomechanical stability are greatly enhanced by Zn/Mg-PIII implants, which may have the potential application in dental implantation in the future. We believe this paper may be of particular interest to the readers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7653
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
C6H4NNa2O6P