EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Anticancer copper pyridine benzimidazole complexes: ROS generation, biomolecule interactions, and cytotoxicity.
PMID 27915178
Abstract
The Cu(II) complex CuCl
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209538
4-Fluorobenzyl bromide, 97%
C7H6BrF
B17905
Benzyl bromide, reagent grade, 98%
C7H7Br