EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Synthesis of nano-hydroxyapatite sorbent for microextraction in packed syringe of phthalate esters in water samples.
PMID 27916131
Abstract
An easy alkoxide-based sol-gel method using Ca(NO3)2·4H2O and tri-ethyl phosphate (PO(OC2H5)3; TEP) precursors employed to synthesize nano-hydroxyapatite (HAP). The structural characterization of samples was studied using XRD, TEM and elemental analysis of SEM. Scherrer and Williamson-Hall equations were applied to measurement of the crystallite size distributions and micro-strain of the synthesized HAP powder. The synthesized HAP nanoparticles were applied as an efficient sorbent for the microextraction in packed syringe (MEPS) of phthalate esters (PAEs) from water samples. Separation and determination of the PAEs were performed by gas chromatography-flame ionization detection (GC-FID). Several variables affecting the extraction efficiency of the analytes i.e., desorption solvent, volume of desorption solvent and number of extraction cycles were investigated and optimized. Under optimized conditions, the calibration curves were linear (R > 0.9916) in the concentration range of 0.05-100 ng mL-1. The method displays detection limits (at an S/N ration of 3) in the range from 0.02 to 0.1 ng mL-1, and the limits of quantification (at an S/N ratio of 10) are between 0.07 and 0.25 ng mL-1. Relative standard deviations (RSDs) for intra- and inter-day precision are 4.8-8.3%, and 6.1-8.5%, respectively. The applicability of the developed method was examined by analyzing different water samples (river water, bottled mineral water and tap water) and the relative recovery values for the spiked water samples were found to be in the range of 85.5-99.2%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274623
Dess-Martin periodinane, 97%
C13H13IO8
E50604
Ethyltriphenylphosphonium bromide, 99%
C20H20BrP