EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Optical properties of secondary organic aerosols derived from long-chain alkanes under various NO
PMID 27916309
Abstract
Long-chain alkanes are a type of important intermediate-volatile organic compounds (IVOCs) in the atmosphere, which contribute to a large proportion of secondary organic aerosol (SOA). However, the optical properties of SOA derived from long-chain alkanes remain poorly understood. Here, we investigate the refractive index (RI) of SOA derived from photo-oxidation of dodecane (C12), pentadecane (C15) and heptadecane (C17) under low-NO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4562
5-(N,N-Dimethyl)amiloride hydrochloride
C8H12ClN7O · HCl