EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
System maps for retention of small neutral compounds on a biphenylsiloxane-bonded silica stationary phase in reversed-phase liquid chromatography.
PMID 27916389
Abstract
The system constants of the solvation parameter model are used to prepare system maps for the retention of small neutral compounds on a biphenylsiloxane-bonded superficially porous silica stationary phase (Kinetex Biphenyl) for aqueous-organic solvent mobile phases containing 10-70% (v/v) methanol or acetonitrile. The retention properties of the biphenylsiloxane-bonded phase are shown to be complementary to an octadecylsiloxane-bonded silica (Kinetex C-18) and a pentafluorophenylpropylsiloxane-bonded silica stationary phases (Discovery HS F5). The retention properties of the Kinetex Biphenyl column are similar to an ether-linked phenylpropylsiloxane-bonded silica phase (Synergi Polar RP) with only small differences in relative retention.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
567501-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 3.3 cm × 2.1 mm
567500-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm
567502-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm
567503-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm
567542-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 3 mm, pkg of 1 ea
567530-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4 mm
567531-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 4 mm
567532-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4 mm
567504-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm
567506-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 4.6 mm
567507-U Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
567508-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
567510-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
567511-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
567512-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
567535-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
567536-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
567513-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
567515-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
567516-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
567517-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
567520-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
567523-U Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
567570-U Discovery® HS F5 Supelguard Cartridge, 3 μm particle size, L × I.D. 2 cm × 2.1 mm
567571-U Discovery® HS F5 Supelguard Cartridge, 3 μm particle size, L × I.D. 2 cm × 2.1 mm
567572-U Discovery® HS F5 Supelguard Cartridge, 3 μm particle size, L × I.D. 2 cm × 4 mm
567573-U Discovery® HS F5 Supelguard Cartridge, 3 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 1 kit
567574-U Discovery® HS F5 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
567575-U Discovery® HS F5 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 1 kit
567576-U Discovery® HS F5 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm