EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Insight into the biochemical, kinetic and spectroscopic characterization of garlic (Allium sativum) phytocystatin: Implication for cardiovascular disease.
PMID 27916569
Abstract
Phytocystatins are cysteine proteinase inhibitors present in plants. They play crucial role in maintaining protease-anti protease balance and are involved in various endogenous processes. Thus, they are suitable and convenient targets for genetic engineering which makes their isolation and characterisation from different sources the need of the hour. In the present study a phytocystatin has been isolated from garlic (Allium sativum) by a simple two-step process using ammonium sulphate fractionation and gel filtration chromatography on Sephacryl S-100HR with a fold purification of 152.6 and yield 48.9%. A single band on native gel electrophoresis confirms the homogeneity of the purified inhibitor. The molecular weight of the purified inhibitor was found to be 12.5kDa as determined by SDS-PAGE and gel filtration chromatography. The garlic phytocystatin was found to be stable under broad range of pH (6-8) and temperature (30°C-60°C). Kinetic studies suggests that garlic phytocystatins are reversible and non-competitive inhibitors having highest affinity for papain followed by ficin and bromelain. UV and fluorescence spectroscopy revealed significant conformational change upon garlic phytocystatin-papain complex formation. Secondary structure analysis was performed using CD and FTIR. Garlic phytocystatin possesses 33.9% alpha-helical content as assessed by CD spectroscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S100HR Sephacryl®, 100-HR, MW range 1000-100,000 Da (globular proteins)