EMAIL THIS PAGE TO A FRIEND
Scientific reports
Inhibition of alpha-synuclein aggregation by multifunctional dopamine agonists assessed by a novel in vitro assay and an in vivo Drosophila synucleinopathy model.
PMID 27917933
Abstract
Aggregation of alpha synuclein (α-syn) leading to dopaminergic neuronal death has been recognized as one of the main pathogenic factors in the initiation and progression of Parkinson's disease (PD). Consequently, α-syn has been targeted for the development of therapeutics for PD. We have developed a novel assay to screen compounds with α-syn modulating properties by mimicking recent findings from in vivo animal studies involving intrastriatal administration of pre-formed fibrils in mice, resulting in increased α-syn pathology accompanying the formation of Lewy-body (LB) type inclusions. We found that in vitro generated α-syn pre-formed fibrils induce seeding of α-syn monomers to produce aggregates in a dose-and time-dependent manner under static conditions in vitro. These aggregates were toxic towards rat pheochromocytoma cells (PC12). Our novel multifunctional dopamine agonists D-519 and D-520 exhibited significant neuroprotection in this assay, while their parent molecules did not. The neuroprotective properties of our compounds were further evaluated in a Drosophila model of synucleinopathy. Both of our compounds showed protective properties in fly eyes against the toxicity caused by α-syn. Thus, our in vitro results on modulation of aggregation and toxicity of α-syn by our novel assay were further validated with the in vivo experiments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3516
Thioflavin T, used as stain for amyloid
C17H19ClN2S