EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Lin28 is induced in primed embryonic stem cells and regulates let-7-independent events.
PMID 27920151
Abstract
Lin28 RNA-binding proteins play important roles in pluripotent stem cells, but the regulation of their expression and the mechanisms underlying their functions are still not definitively understood. Here we address the above-mentioned issues in the first steps of mouse embryonic stem cell (ESC) differentiation. We observed that the expression of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU196671 MISSION® esiRNA, esiRNA targeting mouse Hmga2