EMAIL THIS PAGE TO A FRIEND
Drug design, development and therapy
Design, synthesis and evaluation of VEGF-siRNA/CRS as a novel vector for gene delivery.
PMID 27920500
Abstract
Small interfering RNA (siRNA) delivery is a prospective method in gene therapy, but it has application limitations such as negative charge, water solubility and high molecular weight. In this study, a safe and efficient nano-vector, CRS, was designed and synthesized to facilitate siRNA delivery. Physical and chemical properties of VEGF-siRNA/CRS were characterized by methods including scanning electron microscopy (SEM), transmission electron microscopy, zeta potential (