EMAIL THIS PAGE TO A FRIEND
Biological chemistry
Intra- or extra-exosomal secretion of HDGF isoforms: the extraordinary function of the HDGF-A N-terminal peptide.
PMID 27926477
Abstract
Hepatoma-derived growth factor (HDGF) is a protein with diverse intracellular functions. Moreover, after non-conventional secretion, extracellular HDGF is able to influence different signaling pathways, leading for example to induction of processes like epithelial-mesenchymal transition (EMT) and cell migration. Intriguingly, in recent proteome studies, HDGF was also found secreted by special microvesicles called exosomes. Recently, we demonstrated the existence of two new HDGF isoforms (B and C). These isoforms are involved in different cellular processes than HDGF-A. Along this line, in the present study we discovered that full length HDGF-A clearly is located inside of exosomes, whereas the isoforms HDGF-B and HDGF-C are found exclusively on the outer surface. Furthermore, while HDGF-B and HDGF-C seem to use exosomes mediated pathway exclusively, HDGF-A was found also as unbound protein in the conditioned media. The new finding of an intra- or extra-exosomal localisation of protein splice variants opens a fascinating new perspective concerning functional diversity of HDGF isoforms. Dysregulation of HDGF expression during cancer development and tumor progression is a commonly known fact. With our new findings, unraveling the potential functional impact according to physiological versus pathophysiologically altered levels and compositions of intra- and extra-exosomal HDGF has to be addressed in future studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4562
5-(N,N-Dimethyl)amiloride hydrochloride
C8H12ClN7O · HCl
M1404
Nocodazole, ≥99% (TLC), powder
C14H11N3O3S