EMAIL THIS PAGE TO A FRIEND
Preventing chronic disease
Strengthening the Connection Between the Supplemental Nutrition Assistance Program and Farmer's Markets.
PMID 27930285
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3398
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
C7H12N2O4S