EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
miR-200c enhances sensitivity of drug-resistant non-small cell lung cancer to gefitinib by suppression of PI3K/Akt signaling pathway and inhibites cell migration via targeting ZEB1.
PMID 27930974
Abstract
Acquired resistance to epidermal growth factor receptor tyrosine kinase inhibitors (EGFR-TKIs) is a major obstacle in the treatment of non-small cell lung cancer (NSCLC) patients. We explored the role of miR-200c in modulating the sensitivity of gefitinib-resistant NSCLC cells and examined the underlying mechanism. The gefitinib-resistant cell line PC-9-ZD and its parental PC-9 cells were used. Growth inhibition was detected by MTT assay. The cell apoptosis was detected by Annexin V/PI assay. Cell migration was assessed by wound-healing assay. RT-PCR was used to detected levels of miR-200c and ZEB1. The PI3k, Bcl-2, Bax, caspase-3 and ZEB1 protein expression were detected using Western blot analysis, and TUNEL, Immunohistochemistry for xenograft model. PC-9-ZD cells had low level of miR-200c expression compared to its parental PC-9 cells. PC-9-ZD cells with miR-200c transfection were more sensitive to gefitinib treatment. Apoptosis induced by gefitinib was observed in PC-9-ZD cells with miR-200c transfection significantly. The levels of phosphorylated-Akt and Bcl-2 expression decreased and levels of Bax and Caspase-3 expression increased in PC-9-ZD cells with miR-200c transfection. Cell migration was inhibited and ZEB1 mRNA level and protein expression were significantly decreased in PC-9-ZD cells with miR-200c transfection. Further in gefitinib resistant xenograft model, miR-200c enhanced sensitivity of gefitinib and induced apoptosis significantly through PI3K/Akt signaling pathway and targeting ZEB1. These results provided insights into the functions of miR-200c and offered an alternate approach in treating gefitinib-resistance NSCLC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU151901 MISSION® esiRNA, esiRNA human ZEB1 (esiRNA1)