EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Protein
PMID 27932460
Abstract
Disruption of the
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U4375
Uridine 5′-diphospho-N-acetylglucosamine sodium salt, ≥98%
C17H25N3Na2O17P2