EMAIL THIS PAGE TO A FRIEND
Oncotarget
Analysis of progression-free survival of first-line tyrosine kinase inhibitors in patients with non-small cell lung cancer harboring leu858Arg or exon 19 deletions.
PMID 27935868
Abstract
Gefitinib, erlotinib and afatinib provide remarkable response rates and progression-free survival compared to platinum-based chemotherapy in patients with non-small cell lung cancer harboring epidermal growth factor receptor-activating mutations, and are therefore standard first-line treatment in these patients. However, no study has compared these drugs regarding progression-free survival. We conducted this retrospective study at a single medical center in Taiwan from February 16, 2011 to October 30, 2015. We used the Kaplan-Meier method to estimate survival, and multivariate Cox proportional hazard models to estimate adjusted hazard ratios and 95% confidence intervals. Of the 1006 patients diagnosed with stage IIIb and IV non-small cell lung cancer in the study period, 448 (44.5%) had EGFR-activating mutations and received first-line therapy with gefitinib (n = 304, 67.6%), erlotinib (n = 63, 14.3%), or afatinib (n = 81, 18.1%). The median duration of follow-up for progression-free survival was 12.1 months in the gefitinib arm (Interquartile range [IQR]: 5.5-16.5), 11.2 months in the erlotinib arm (IQR: 4.9-16.7), and 10.3 months in the afatinib arm (IQR: 7.0-14.2). Progression-free survival was significantly longer in the patients who received afatinib or erlotinib compared to those who received gefitinib (log-rank test, p < 0.001), and the median progression-free survival was 11.4 months in the gefitinib group. Afatinib and erlotinib provide significant benefits in progression-free survival compared to gefitinib in first-line treatment of patients with non-small-cell lung cancers harboring EGFR-activating mutations. Further clinical trials are warranted to validate these findings.