EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Isolation of isoflavonoids possessing antioxidant activity from the fermentation broth of Streptomyces sp.
PMID 2793588
Abstract
Three antioxidant isoflavonoids characterized as 4',7,8-trihydroxyisoflavone (1), 3',4',7-trihydroxyisoflavone (2) and 8-chloro-3',4',5,7-tetrahydroxyisoflavone (3) were isolated from the cultured broth of Streptomyces sp. OH-1049. Among them, 3 is a novel isoflavonoid possessing a chlorine atom in the molecule. In in vitro studies, these antibiotics were found to possess antioxidant activity whereas showed almost no cytocidal activities against HeLa S3 cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0947 YN1, ≥98% (HPLC)
C15H10O5