EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Antitumor activity of chrysomycins M and V.
PMID 2793599
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6616
Chrysomycin A, ≥98% (HPLC), solid
C28H28O9
C6491
Chrysomycin B, ≥98% (HPLC), solid
C27H28O9