EMAIL THIS PAGE TO A FRIEND
Planta medica
Relaxant Effect of Sodium Tanshinone IIA Sulphonate on Mouse Tracheal Smooth Muscle.
PMID 27936472
Abstract
Sodium tanshinone IIA sulphonate, a water-soluble derivative of tanshinone IIA, has been proven to possess versatile biological properties, but its pharmacological effect on tracheal smooth muscle remains elusive. This paper presents a study on the relaxant effect and underlying mechanisms of sodium tanshinone IIA sulphonate on mouse tracheal smooth muscle. The relaxant effect of sodium tanshinone IIA sulphonate was evaluated in mouse tracheal rings using a mechanical recording system. Intracellular Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46393
Fluo-3, suitable for fluorescence, ~70%
C36H30Cl2N2O13