EMAIL THIS PAGE TO A FRIEND
Journal of proteome research
Unraveling the Proteome Composition and Immuno-profiling of Western India Russell's Viper Venom for In-Depth Understanding of Its Pharmacological Properties, Clinical Manifestations, and Effective Antivenom Treatment.
PMID 27936776
Abstract
The proteome composition of western India (WI) Russell's viper venom (RVV) was correlated with pharmacological properties and pathological manifestations of RV envenomation. Proteins in the 5-19 and 100-110 kDa mass ranges were the most predominate (∼35.1%) and least abundant (∼3.4%) components, respectively, of WI RVV. Non-reduced SDS-PAGE indicated the occurrence of multiple subunits, non-covalent oligomers, self-aggregation, and/or interactions among the RVV proteins. A total of 55 proteins belonging to 13 distinct snake venom families were unambiguously identified by ESI-LC-MS/MS analysis. Phospholipase A