EMAIL THIS PAGE TO A FRIEND
Metabolic engineering
Integration of Biomass Formulations of Genome-Scale Metabolic Models with Experimental Data Reveals Universally Essential Cofactors in Prokaryotes.
PMID 27939572
Abstract
The composition of a cell in terms of macromolecular building blocks and other organic molecules underlies the metabolic needs and capabilities of a species. Although some core biomass components such as nucleic acids and proteins are evident for most species, the essentiality of the pool of other organic molecules, especially cofactors and prosthetic groups, is yet unclear. Here we integrate biomass compositions from 71 manually curated genome-scale models, 33 large-scale gene essentiality datasets, enzyme-cofactor association data and a vast array of publications, revealing universally essential cofactors for prokaryotic metabolism and also others that are specific for phylogenetic branches or metabolic modes. Our results revise predictions of essential genes in Klebsiella pneumoniae and identify missing biosynthetic pathways in models of Mycobacterium tuberculosis. This work provides fundamental insights into the essentiality of organic cofactors and has implications for minimal cell studies as well as for modeling genotype-phenotype relations in prokaryotic metabolic networks.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
82870
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
C8H10NO6P · H2O