The international journal of biochemistry & cell biology
Increasing NO level regulates apoptosis and inflammation in macrophages after 2-chloroethyl ethyl sulphide challenge.
PMID 27939754
Generation of nitric oxide (NO) in cellular compartments acts in a redox-dependent manner to counteract oxidative stress either by directly acting as an antioxidant through scavenging superoxide anions (O2(-)), to form peroxynitrite (ONOO-) or acting as a signaling molecule, altering gene expression that triggers various physiological processes. However, the molecular mechanisms of macrophage activation and NO production leads to apoptosis and inflammation after 2-chloroethyl ethyl sulphide (CEES) exposure remains unclear. We showed that CEES exposure in macrophages increased the O2(-) production. Also CEES exposure transiently increases the NO production and ONOO- accumulation via expression of inducible NO synthase (iNOS). Simultaneously, CEES exposure caused a significant reduction in cellular antioxidants and modulate lipid peroxidation (LPO), and protein carbonylation (PC) reactions, which was correlated with the increased level of NO and ONOO- accumulation. Mechanistic studies showed the DNA damage, 8-oxoGDNA glycosylase (OGG1) down regulation and 8-hydroxydeoxyguanosine (8-OHdG) accumulations in DNA, which was also confirmed by phosphorylation of ATM, ATR and H2A.X. Elevated levels of NO/ONOO- plays an important role in apoptosis, and alteration of cell cycle regulatory proteins in macrophages after CEES exposure. Moreover, CEES exposure to macrophage cells enhanced the transcriptional activities of inflammatory mediators such as TNFα, IL-1α, ICAM, CX3CL1, CCL8, and CXCL10, which were linked with NO/ONOO- accumulation. These results showed a mechanistic explanation of how NO/ONOO- cooperate to conduct apoptosis and inflammatory signals in macrophages after CEES challenged. Further, the protective effects of NO/ONOO- inhibitors may provide the basis for the development of a therapeutic strategy to counteract exposure to CEES.
Related Materials
Product #
Molecular Formula
Add to Cart
Dihydrorhodamine 123, ≥95%