EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Electrospun manganese (III) oxide nanofiber based electrochemical DNA-nanobiosensor for zeptomolar detection of dengue consensus primer.
PMID 27940241
Abstract
Nanoscale biosensors, owing to their high-sensitivity and extremely low limits-of-detection, have enabled the realization of highly complex and sophisticated miniaturized platforms for several important healthcare applications, the most predominant one being disease diagnosis. In particular, nanomaterial facilitated electrochemical detection of DNA hybridization has had an exceptional impact on fields such as genetics and cancerous mutation detection Here we report an ultrasensitive electrochemical platform using electrospun semi-conducting Manganese (III) Oxide (Mn