EMAIL THIS PAGE TO A FRIEND
Cardiovascular pathology : the official journal of the Society for Cardiovascular Pathology
CaMKIIδ meditates phenylephrine induced cardiomyocyte hypertrophy through store-operated Ca
PMID 27940402
Abstract
Evidence suggests that store-operated Ca2+ entry (SOCE) is involved in the hypertrophy of cardiomyocytes. The signaling mechanisms of SOCE contributing to cardiac hypertrophy following phenylephrine (PE) stimulation are not fully understood. Ca