EMAIL THIS PAGE TO A FRIEND
Genes
Detection and Quantification of the Fragile X Mental Retardation Protein 1 (FMRP).
PMID 27941672
Abstract
The final product of FMR1 gene transcription, Fragile X Mental Retardation Protein 1 (FMRP), is an RNA binding protein that acts as a repressor of translation. FMRP is expressed in several tissues and plays important roles in neurogenesis, synaptic plasticity, and ovarian functions and has been implicated in a number of neuropsychological disorders. The loss of FMRP causes Fragile X Syndrome (FXS). In most cases, FXS is due to large expansions of a CGG repeat in FMR1-normally containing 6-54 repeats-to over 200 CGGs and identified as full mutation (FM). Hypermethylation of the repeat induces FMR1 silencing and lack of FMRP expression in FM male. Mosaic FM males express low levels of FMRP and present a less severe phenotype that inversely correlates with FMRP levels. Carriers of pre-mutations (55-200 CGG) show increased mRNA, and normal to reduced FMRP levels. Alternative splicing of FMR1 mRNA results in 24 FMRP predicted isoforms whose expression are tissues and developmentally regulated. Here, we summarize the approaches used by several laboratories including our own to (a) detect and estimate the amount of FMRP in different tissues, developmental stages and various pathologies; and (b) to accurately quantifying FMRP for a direct diagnosis of FXS in adults and newborns.