EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
An absorption mechanism and polarity-induced viscosity model for CO
PMID 27942645
Abstract
A series of new hydroxypyridine-based ionic liquids (ILs) are synthesized and applied in CO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
438294
Tetrabutylphosphonium hydroxide solution, 40 wt. % in H2O
C16H37OP