EMAIL THIS PAGE TO A FRIEND
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology
Mathematical description of pH-stat kinetic traces measured during photochemical quinone decomposition.
PMID 27942682
Abstract
Substituted 1,4-benzoquinone (QR) derivatives are photosensitive in aqueous solution and form hydroquinones (QR-H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
431974
2,5-Dichloro-1,4-benzoquinone, 98%
C6H2Cl2O2