EMAIL THIS PAGE TO A FRIEND
Molecular & cellular proteomics : MCP
Structural Analysis of Variable Domain Glycosylation of Anti-Citrullinated Protein Antibodies in Rheumatoid Arthritis Reveals the Presence of Highly Sialylated Glycans.
PMID 27956708
Abstract
Recently, we showed the unexpectedly high abundance of N-linked glycans on the Fab-domain of Anti-Citrullinated Protein Antibodies (ACPA). As N-linked glycans can mediate a variety of biological functions, we now aimed at investigating the structural composition of the Fab-glycans of ACPA-IgG to better understand their mediated biological effects. ACPA-IgG and noncitrulline specific (control) IgG from plasma and/or synovial fluid of nine ACPA positive rheumatoid arthritis patients were affinity purified. The N-linked glycosylation of total, Fc and F(ab')