EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Chemopreventive Potential of Powdered Red Wine Pomace Seasonings against Colorectal Cancer in HT-29 Cells.
PMID 27957845
Abstract
This study evaluates the antiproliferative and antigenotoxic actions of powdered red wine pomace seasonings (Sk-S, seedless; W-S, whole; Sd-S, seeds). In vitro gastrointestinal digested and colonic fermented fractions of the seasonings were used as cell treatments. Phenolic acids from Sk-S showed the highest bioaccessibility in the small intestine, whereas polyphenols contained in Sd-S might be the most fermentable in the colon. Dietary fiber from Sk-S was the best substrate for short chain fatty acids production by gut microbiota. Colon cancerous (HT-29) cell viability was inhibited by 50% (IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48640
Ethyl gallate, antioxidant, ≥96.0% (HPLC)
C9H10O5