EMAIL THIS PAGE TO A FRIEND
Biotechnology and applied biochemistry
Degradation of phytosterols in tobacco waste extract by a novel Paenibacillus sp.
PMID 27958658
Abstract
Phytosterols have been demonstrated to be precursors of polycyclic aromatic hydrocarbons (PAHs) formed during biomass pyrolysis. Here, a novel Paenibacillus sp. was evaluated for its ability to degrade phytosterols in tobacco waste extract (TWE). The optimal conditions for cell growth and stigmasterol (a representative of phytosterols) degradation were 37 °C, pH 7.0, 1.0 g/L yeast extract, and 6.0 g/L glucose. Paenibacillus sp. could degrade stigmasterol under high concentrations of glucose (up to 130 g/L) and tolerate wide pH (5.0-9.0) and temperature (25-42 °C) ranges. The new strain could degrade stigmasterol completely into CO